(4-Bromophenyl)acetic acid ethyl ester 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate
392990010 392990050 392990250
ACRO392990010EA 240 SEK
ACRO392990010 ACRO392990050 ACRO392990250
(4-Bromophenyl)acetic acid ethyl ester 99%
(4-Bromophenyl)acetic acid ethyl ester
Formel: C₁₀H₁₁BrO₂
Smältpunkt: 29…31 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016333
CAS nummer: 14062-25-0
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥98.5 %