(4-(Bromomethyl)phenyl)acetic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-(Bromomethyl)phenylacetic acid
392900010 392900050
ACRO392900010EA 785 SEK
ACRO392900010 ACRO392900050
(4-(Bromomethyl)phenyl)acetic acid 97%
(4-(Bromomethyl)phenyl)acetic acid
Formel: (C₆H₄(CH₂Br))CH₂COOH
MW: 229,07 g/mol
Smältpunkt: 180…182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010632
CAS nummer: 13737-36-5
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %