α-Bromo-3-iodotoulene 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide
392530010 392530050 392530250
ACRO392530010EA 800 SEK
ACRO392530010 ACRO392530050 ACRO392530250
α-Bromo-3-iodotoulene 97%
α-Bromo-3-iodotoulene
Formel: C₇H₆BrI
MW: 296.93 g/mol
Smältpunkt: 47…50 °C
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00019016
CAS nummer: 49617-83-6
UN: 3261
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Light orange to red to brown Crystalline powder
Infrared spectrum Conforms
Melting point 47 to 50 °C
GC ≥96.0 %