Diethoxymethylsilane 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Diethoxy(methyl)silane
392260050 392260250 392261000 392260000
ACRO392260050EA 740 SEK
ACRO392260050 ACRO392260250 ACRO392261000 392260000.
Diethoxymethylsilane 97%
Diethoxymethylsilane
Formel: C₅H₁₄O₂Si
MW: 134.25 g/mol
Flampunkt: 10 °C
Storage Temperature: Omgivande
MDL Number: MFCD00026749
CAS nummer: 2031-62-1
UN: 1993
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3740 to 1.3760 (20°C, 589 nm)
GC ≥96.0 %