3,5-Di-tert-butylsalicylic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,5-Bis-tert-butylsalicylic acid, 3,5-Di-tert-butyl-2-hydroxybenzoic Acid
386860050 386860250
ACRO386860050EA 520 SEK
ACRO386860050 ACRO386860250
3,5-Di-tert-butylsalicylic acid 99%
3,5-Di-tert-butylsalicylic acid
Appearance: Off-white to beige Powder
Formel: C₁₅H₂₂O₃
MW: 250,34 g/mol
Smältpunkt: 158…162 °C
Flampunkt: 179 °C
Storage Temperature: Omgivande
MDL Number: MFCD00059058
CAS nummer: 19715-19-6
Order Now
Specifikation testresultat
Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 160 to 165 °C
HPLC ≥98.5 %