α-Bromo-4-chlorotoulene 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide
391710050 391710250 391711000
ACRO391710050EA 910 SEK
ACRO391710050 ACRO391710250 ACRO391711000
α-Bromo-4-chlorotoulene 98%
α-Bromo-4-chlorotoulene
Formel: C₇H₆BrCl
MW: 205.48 g/mol
Kokpunkt: 118…123 °C (19 mmHg)
Smältpunkt: 46…50 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040714
CAS nummer: 622-95-7
UN: 3261
ADR: 8 II
Order Now
Specifikation testresultat
Appearance White to almost white Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %