1-Chloroisoquinoline 97+%
Leverantör: Thermo Scientific
Varning
391380010 391380050
ACRO391380010EA 815 SEK
ACRO391380010 ACRO391380050
1-Chloroisoquinoline 97+%
1-Chloroisoquinoline
Formel: C₉H₆ClN
MW: 163,61 g/mol
Smältpunkt: 31…36 °C
Storage Temperature: Omgivande
MDL Number: MFCD00024134
CAS nummer: 19493-44-8
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to yellow Crystals and/or chunks or low melting solid
Infrared spectrum Conforms
GC ≥97.0 %