2-Chloroisonicotinic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid
397790050 397790250
ACRO397790050EA 1000 SEK
ACRO397790050 ACRO397790250
2-Chloroisonicotinic acid 97%
2-Chloroisonicotinic acid
Formel: C₆H₄ClNO₂
MW: 157,56 g/mol
Smältpunkt: 223 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191402
CAS nummer: 6313-54-8
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %