1.0 M i vatten
Leverantör: Thermo Fisher Scientific
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
397080010 397080025
ACRO397080010EA 1175 SEK
ACRO397080010 ACRO397080025
1.0 M i vatten
Dikaliumhydrogenfosfat
Formel: K₂HPO₄
MW: 174.18 g/mol
Smältpunkt: 340
Storage Temperature: Omgivande
MDL Number: MFCD00011383
CAS nummer: 7758-11-4
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Solution
Titration with HClO4 1.0 M Exact strength on the certificate of analysis