tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-BOC-3-pyrrolidinone
397050010 397050100
ACRO397050010EA 1075 SEK
ACRO397050010 ACRO397050100
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate
Appearance: White to light brown Low melting solid
Formel: C₉H₁₅NO₃
MW: 185,22 g/mol
Smältpunkt: 34…38 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01631194
CAS nummer: 101385-93-7
Order Now
Specifikation testresultat
Appearance White to light brown Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %