4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride
397040010 397040050
ACRO397040010EA 395 SEK
ACRO397040010 ACRO397040050
4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride
Formel: C₇H₈ClF₃N₂O
MW: 228.60 g/mol
Smältpunkt: 230 °C
Storage Temperature: Omgivande
MDL Number: MFCD00053033
CAS nummer: 133115-72-7
Order Now
Specifikation testresultat
Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %