3-(Boc-amino)propylbromide 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-(tert-Butoxycarbonylamino)propyl bromide, 3-(Boc-amino)propyl bromide, tert-Butyl N-(3-Bromopropyl)carbamate, N-(3-Bromopropyl)carbamic acid tert-Butyl ester
397010010 397010050
ACRO397010010EA 3000 SEK
ACRO397010010 ACRO397010050
3-(Boc-amino)propylbromide 95%
3-(Boc-amino)propylbromide
Appearance: White Low melting solid
Formel: C8H16BrNO2
MW: 238,1221 g/mol
Smältpunkt: 34…36,5 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02683429
CAS nummer: 83948-53-2
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White Low melting solid
Infrared spectrum Conforms
GC ≥94 %