1-Boc-3-piperidone 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-BOC-3-piperidone, 1-Boc-3-Piperidinone
397000050 397000250 397001000
ACRO397000050EA 1625 SEK
ACRO397000050 ACRO397000250 ACRO397001000
1-Boc-3-piperidone 97%
1-Boc-3-piperidone
Formel: C₁₀H₁₇NO₃
MW: 199,25 g/mol
Smältpunkt: 37…39 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631193
CAS nummer: 98977-36-7
Order Now
Specifikation testresultat
Appearance White to yellow to orange Solid or liquid
Infrared spectrum Conforms
GC ≥96.0 %