1-Methylcyclopropanecarboxylic acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-methylcyclopropane-1-carboxylic acid
194960010 194960050
ACRO194960010EA 1125 SEK
ACRO194960010 ACRO194960050
1-Methylcyclopropanecarboxylic acid 98%
1-Methylcyclopropanecarboxylic acid
Formel: C₅H₈O₂
MW: 100,12 g/mol
Smältpunkt: 34…36 °C
Flampunkt: 84 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001290
CAS nummer: 6914-76-7
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %