Pyrrole-2-carboxylic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
132070010 132070050 132070250
ACRO132070010EA 790 SEK
ACRO132070010 ACRO132070050 ACRO132070250
Pyrrole-2-carboxylic acid 97%
Pyrrole-2-carboxylic acid
Formel: C₅H₅NO₂
MW: 111.10 g/mol
Smältpunkt: 207…208 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005219
CAS nummer: 634-97-9
Order Now
Specifikation testresultat
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure