Cyclohexanemethylamine 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine
146570050 146570250
ACRO146570050EA 1100 SEK
ACRO146570050 ACRO146570250
Cyclohexanemethylamine 97%
Cyclohexanemethylamine
Formel: C₇H₁₅N
MW: 113.2 g/mol
Kokpunkt: 161…163 °C (1013 hPa)
Smältpunkt: –8 °C
Densitet: 0.87 g/cm³
Flampunkt: 43 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001520
CAS nummer: 3218-02-8
UN: 2734
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥97.5 %