Cycloheptanecarboxylic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
111020050 111020250
ACRO111020050EA 1900 SEK
ACRO111020050 ACRO111020250
Cycloheptanecarboxylic acid 97%
Cycloheptanecarboxylic acid
Formel: C₈H₁₄O₂
Kokpunkt: 135…138 °C (9 mmHg)
Smältpunkt: 71 °C
Densitet: 1,035 g/cm³
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004152
CAS nummer: 1460-16-8
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4730 (20°C, 589 nm)
Titration with NaOH ≥96.0 %