2-Nitro-1-phenylpropene 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: trans-β-Methyl-β-nitrostyrene
386510050 386510010 386510250
ACRO386510050EA 1100 SEK
ACRO386510050 ACRO386510010 ACRO386510250
2-Nitro-1-phenylpropene 99%
2-Nitro-1-phenylpropene
Formel: C₉H₉NO₂
Smältpunkt: 64…67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014720
CAS nummer: 705-60-2
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 67 °C
GC ≥98.5 %