Cyclopentanecarbaldehyde 97% stabilised

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: Cyclopentanecarboxaldehyde

425910050 425910010 425910250
ACRO425910050EA 2650 SEK
ACRO425910050 ACRO425910010 ACRO425910250
Cyclopentanecarbaldehyde 97% stabilised
Cyclopentanecarbaldehyde
Formel: C₆H₁₀O
MW: 98.14 g/mol
Kokpunkt: 140…141 °C (760 mmHg)
Densitet: 0.919 g/cm³
Flampunkt: 28 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00060798
CAS nummer: 872-53-7
UN: 1989
ADR: 3 III

Order Now

Specifikation testresultat

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4480 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.1 % w/w hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR