(2-Bromoethoxy)-tert-butyldimethylsilane 98+% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: (2-Bromoethoxy)(tert-butyl)dimethylsilane
399830100 399830010
ACRO399830100EA 3300 SEK
ACRO399830100 ACRO399830010
(2-Bromoethoxy)-tert-butyldimethylsilane 98+% stabilised
(2-Bromoethoxy)-tert-butyldimethylsilane
Appearance: Colorless Liquid
Formel: C₈H₁₉BrOSi
MW: 239,228 g/mol
Kokpunkt: 70…75 °C (2,5 mmHg)
Densitet: 1,115 g/cm³
Flampunkt: 73 °C
Storage Temperature: Omgivande
MDL Number: MFCD00209550
CAS nummer: 86864-60-0
Order Now
Specifikation testresultat
Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥98.0 %
Stabilizer ≤5 % Na2CO3