Methyl-3-aminopropanoate hydrochloride 98%
Leverantör: Thermo Scientific
Synonyms: beta-Alanine methyl ester hydrochloride, H-β-Ala-OMe·HCl, 3-Aminopropionic Acid Methyl Ester Hydrochloride, Methyl 3-aminopropanoate hydrochloride, Methyl 3-Aminopropionate Hydrochloride, H-β-Ala-OMe.HCl
371470050 371470250
ACRO371470050EA 850 SEK
ACRO371470050 ACRO371470250
Methyl-3-aminopropanoate hydrochloride 98%
Methyl-3-aminopropanoate hydrochloride
CAS No.: 3196-73-4
Formel: C₄H₉NO₂·HCl
MW: 139,6 g/mol
Smältpunkt: 103…105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00039060
CAS nummer: 3196-73-4
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %