(1S,3R)-3-Aminocyclopentanecarboxylic acid 95%, > 90% ee
Leverantör: Thermo Scientific
Varning
362280010 362280050
ACRO362280010EA 5400 SEK
ACRO362280010 ACRO362280050
(1S,3R)-3-Aminocyclopentanecarboxylic acid 95%, > 90% ee
(1S,3R)-3-Aminocyclopentanecarboxylic acid
CAS No.: 71830-07-4
Formel: C₆H₁₁NO₂
Smältpunkt: 192 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00211290
CAS nummer: 71830-07-4
Order Now
Specifikation testresultat
Appearance White to beige Powder
Infrared spectrum Conforms
Enantiomeric excess ≥90 %
Specific optical rotation +5° to +9° (20°C, 589 nm) (C=1, Water)
Titration with HClO4 ≥94 %