5-(4-Chlorophenyl)-2-furoic acid 95+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5-(4-Chlorophenyl)furan-2-carboxylic acid
375540010 375540050
ACRO375540010EA 875 SEK
ACRO375540010 ACRO375540050
5-(4-Chlorophenyl)-2-furoic acid 95+%
5-(4-Chlorophenyl)-2-furoic acid
CAS No.: 41019-45-8
Formel: C₁₁H₇ClO₃
MW: 222.624 g/mol
Smältpunkt: 198…201 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00216587
CAS nummer: 41019-45-8
Order Now
Specifikation testresultat
Appearance Light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥95.0 %