Tributyltin trifluoromethanesulphonate 96%
Leverantör: Thermo Fisher Scientific
Fara
388930010 388930050
ACRO388930010EA 610 SEK
ACRO388930010 ACRO388930050
Tributyltin trifluoromethanesulphonate 96%
Tributyltin trifluoromethanesulphonate
Appearance: White to light beige Crystals and/or chunks
Formel: C₁₃H₂₇F₃O₃SSn
Kokpunkt: 56…60 °C (25 mmHg)
Smältpunkt: 31 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00043324
CAS nummer: 68725-14-4
UN: 2923
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to light beige Crystals and/or chunks
Infrared spectrum Conforms
Titration with NaOH ≥95 %