Thiochroman-4-one 98%
Leverantör: Thermo Fisher Scientific
153480050 153480100
ACRO153480050EA 1200 SEK
ACRO153480050 ACRO153480100
Thiochroman-4-one 98%
Thiochroman-4-one
Appearance: Orange Low melting solid
Formel: C₉H₈OS
MW: 164.222 g/mol
Kokpunkt: 154 °C (12 mmHg)
Smältpunkt: 28…30 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006882
CAS nummer: 3528-17-4
UN: 3335
ADR: 9 III
Order Now
Specifikation testresultat
Appearance Orange Low melting solid
Infrared spectrum Conforms
GC ≤5 % (methanol)