Vinylene carbonate 98% Stabilised
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1,3-Dioxol-2-one
140890050 140890250
ACRO140890050EA 1250 SEK
ACRO140890050 ACRO140890250
Vinylene carbonate 98% Stabilised
Vinylene carbonate
Appearance: Clear colorless to light yellow Liquid or solid
Formel: C₃H₂O₃
MW: 86.05 g/mol
Kokpunkt: 162 °C (760 mmHg)
Smältpunkt: 19…22 °C
Densitet: 1.36 g/cm³
Flampunkt: 73 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005380
CAS nummer: 872-36-6
UN: 2810
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4230 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer ≤0.1 % BHT