Hematein
Leverantör: Thermo Fisher Scientific
Synonyms: Hydroxybrazilein, Hydroxybrasilein
370840050 370840250
ACRO370840050EA 1500 SEK
ACRO370840050 ACRO370840250
Hematein
Hematein
CAS No.: 475-25-2
Formel: C₁₆H₁₂O₆
MW: 300.266 g/mol
Smältpunkt: 180 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036187
CAS nummer: 475-25-2
Order Now
Specifikation testresultat
Appearance Dark brown Crystalline powder
Infrared spectrum Conforms
Moisture ≤15 %
UV spectrophotometry 5 to 20 %