D-(-)-Pantolactone 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Pantolactone, (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, D-(-)-Pantoyl Lactone, D-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone
218140250 218140050 218141000
ACRO218140250EA 1325 SEK
ACRO218140250 ACRO218140050 ACRO218141000
D-(-)-Pantolactone 99%
D-(-)-Pantolactone
CAS No.: 599-04-2
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Kokpunkt: 120…122 °C (15 mmHg)
Smältpunkt: 88…92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005392
CAS nummer: 599-04-2
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥98.5 %
Specific optical rotation -49° to -52° (25 °C, 589 nm) (c=2, H2O)