Phenyl phosphate, disodium salt dihydrate 98%
Leverantör: Thermo Scientific
Synonyms: Sodium phenyl phosphate dibasic dihydrate
211830100 211831000
ACRO211830100EA 695 SEK
ACRO211830100 ACRO211831000
Phenyl phosphate, disodium salt dihydrate 98%
Phenyl phosphate, disodium salt dihydrate
Appearance: White to off-white Crystalline powder
Formel: C₆H₉Na₂O₆P
MW: 254,085 g/mol
Smältpunkt: >300 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00149080
CAS nummer: 66778-08-3
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 12 to 16 % (K.F.)
Titration with HClO4 ≥97.5 %