Di(N-succinimidyl) carbonate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate
209600250 209600050
ACRO209600250EA 4075 SEK
ACRO209600250 ACRO209600050
Di(N-succinimidyl) carbonate 98%
Di(N-succinimidyl) carbonate
Appearance: White to light yellow Crystalline powder
Formel: C₉H₈N₂O₇
MW: 256,17 g/mol
Smältpunkt: 190 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009767
CAS nummer: 74124-79-1
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %