Naringin 97%
Leverantör: Thermo Scientific
Synonyms: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-rhamnoglucoside
206910250 206911000
ACRO206910250EA 1225 SEK
ACRO206910250 ACRO206911000
Naringin 97%
Naringin
CAS No.: 10236-47-2
Formel: C₂₇H₃₂O₁₄
MW: 580,53 g/mol
Smältpunkt: 166 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00148888
CAS nummer: 10236-47-2
Order Now
Specifikation testresultat
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation -77° to -93° (20°C, 589 nm) (c=1, ethanol)