Nickel aluminium alloy according to Raney (Al-Ni) 50/50, pulver
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Aluminum-nickel alloy
206231000 206235000 206230025
206231000.EA 970 SEK
206231000. 206235000. 206230025.
Nickel aluminium alloy according to Raney (Al-Ni) 50/50, pulver
Nickel aluminium alloy according to Raney
CAS No.: 12635-29-9
Formel: Ni:Al;₅₀:₅₀%
Smältpunkt: 1350 °C
Densitet: 3,46 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00167392
CAS nummer: 12635-29-9
UN: 3208
ADR: 4.3,II
Order Now
Specifikation testresultat
Appearance Grey to grey-black Powder
Aluminium (Al) 47.0 to 52.0 %
Nickel (Ni) 48.0 to 53.0 %