DL-2,3-Diaminopropionic acid hydrochloride 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,3-Diaminopropanoic acid hydrochloride, 3-Amino-DL-alanine Hydrochloride, DL-2,3-Diaminopropionic acid monohydrochloride
204670050 204670250
ACRO204670050EA 1950 SEK
ACRO204670050 ACRO204670250
DL-2,3-Diaminopropionic acid hydrochloride 99%
DL-2,3-Diaminopropionic acid hydrochloride
CAS No.: 54897-59-5
Formel: C₃H₈N₂O₂
MW: 140,57 g/mol
Smältpunkt: 222…226 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00012884
CAS nummer: 54897-59-5
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %