Ethyl-3-methyl-2-oxobutyrate 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ethyl 3-methyl-2-oxobutanoate
203800050 203800250
ACRO203800050EA 1050 SEK
ACRO203800050 ACRO203800250
Ethyl-3-methyl-2-oxobutyrate 95%
Ethyl-3-methyl-2-oxobutyrate
Appearance: Clear light yellow Liquid
Formel: C₇H₁₂O₃
Kokpunkt: 62 °C (11 mmHg)
Densitet: 0,98 g/cm³
Flampunkt: 43 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009122
CAS nummer: 20201-24-5
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4130 (20°C, 589 nm)
GC ≥94 %