Amberlyst® 15, wet ion-exchange resin
Leverantör: Thermo Fisher Scientific
Fara
202150250 202155000 202150050
ACRO202150250EA 435 SEK
ACRO202150250 ACRO202155000 ACRO202150050
Amberlyst® 15, wet ion-exchange resin
Amberlyst® 15
Appearance: Grey-brown =<1.70
Storage Temperature: Omgivande
MDL Number: MFCD00145841
CAS nummer: 9037-24-5
Order Now
Specifikation testresultat
Appearance Grey-brown Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.600 to 0.850 mm
Uniformity coefficient ≤1.70
Moisture holding capacity 52.0 to 57.0 % (H)
Total volume capacity ≥1.70 eq/l (H)
Total weight capacity ≥4.70 eq/kg (H)