Amberlite® CG-50 ion-exchange resin Type 1
Leverantör: Thermo Fisher Scientific
202110250 202115000 202111000
ACRO202110250EA 1175 SEK
ACRO202110250 ACRO202115000 ACRO202111000
Amberlite® CG-50 ion-exchange resin Type 1
Amberlite® CG-50
CAS No.: 9042-11-9
Storage Temperature: Omgivande
MDL Number: MFCD00132693
CAS nummer: 9042-11-9
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Particle size ≤10 % (> 150 µm)
Moisture ≤10 %
Total weight capacity ≥10 meq/g