Pararosaniline base
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tris(4-aminophenyl)methanol, Paramagenta, p-Rosaniline, Parafuchsin, Pararosaniline base
201550100 201550500
ACRO201550100EA 1100 SEK
ACRO201550100 ACRO201550500
Pararosaniline base
Pararosaniline
CAS No.: 467-62-9
Formel: C₁₉H₁₉N₃O
MW: 305.38 g/mol
Smältpunkt: 205 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036222
CAS nummer: 467-62-9
Order Now
Specifikation testresultat
Appearance Purple Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies