Osmiumtetraoxid 2,5% (w/w) i tert-butanol stabilised
Leverantör: Thermo Fisher Scientific
Fara
197450050 197450010 197450250
ACRO197450050EA 2725 SEK
ACRO197450050 ACRO197450010 ACRO197450250
Osmiumtetraoxid 2,5% (w/w) i tert-butanol stabilised
Osmiumtetraoxid
Formel: O₄Os
Flampunkt: 4 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00011150
CAS nummer: 20816-12-0
UN: 1992
ADR: 3,II
Order Now
Specifikation testresultat
Appearance White to yellow Fused mass or solution
Stabilizer tert-Butyl hydroperoxide
Strength ≥2.5 wt%