Poly(styrene), M.W. ∼ 260,000
Leverantör: Thermo Scientific
178890010 178890250 178895000 178891000
ACRO178890010EA 6075 SEK
ACRO178890010 ACRO178890250 ACRO178895000 ACRO178891000
Poly(styrene), M.W. ∼ 260,000
Poly(styrene)
CAS No.: 9003-53-6
Formel: C₈H₈
MW: 125000 - 250000 g/mol
Smältpunkt: 93 °C
Densitet: 1,05 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00084450
CAS nummer: 9003-53-6
Order Now
Specifikation testresultat
Appearance Clear Pellets
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %