Sulphuric acid-[D₂] 98% (w/w) i D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Sulphuric acid-[²H₂]
175480500 175481000 175480050
ACRO175480500EA 2775 SEK
ACRO175480500 ACRO175481000 ACRO175480050
Sulphuric acid-[D₂] 98% (w/w) i D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Sulphuric acid-[D₂]
Formel: D₂SO₄
MW: 100.09 g/mol
Densitet: 1.87 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003542
CAS nummer: 13813-19-9
UN: 1830
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear colorless Solution
D-Enrichment ≥99.50 %
Titration with NaOH 97.5 to 102.5 %
GC ≥94.0 %