4-(2-Aminoethyl)-2-methoxyphenol hydrochloride 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Methoxytyramine·HCl, 3-O-Methyldopamine hydrochloride
172850010 172851000
ACRO172850010EA 4675 SEK
ACRO172850010 ACRO172851000
4-(2-Aminoethyl)-2-methoxyphenol hydrochloride 99+%
4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
Appearance: Light beige Crystalline powder
Formel: C₉H₁₄ClNO₂
MW: 203,666 g/mol
Smältpunkt: 213…215 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012902
CAS nummer: 1477-68-5
Order Now
Specifikation testresultat
Appearance Light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %