5-Vinylbicyclo[2.2.1]hept-2-ene 98% stabilised
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
5-Vinyl-2-norbornene
164860250
164865000
164860025
ACRO164860250EA
670
SEK
ACRO164860250
ACRO164865000
ACRO164860025
5-Vinylbicyclo[2.2.1]hept-2-ene 98% stabilised
5-Vinylbicyclo[2.2.1]hept-2-ene
CAS No.: 3048-64-4
Formel:
C₉H₁₂ MW: 120,20 g/mol Kokpunkt: 141 °C (760 mmHg) Smältpunkt: –80 °C Flampunkt: 27 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00080691 CAS nummer: 3048-64-4 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4800 to 1.4820 (20°C, 589 nm) |
GC | ≥97.5 % |
Color scale | ≤50 APHA |
Stabilizer | 100 to 200 ppm t-Butylcatechol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...