Silikonolja, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
Leverantör: Thermo Fisher Scientific
163850010 163855000 163850025
ACRO163850010EA 3700 SEK
ACRO163850010 ACRO163855000 ACRO163850025
Silikonolja, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
Silikonolja
Smältpunkt: -50 °C
Densitet: 0,96
Flampunkt: 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00132673
CAS nummer: 68083-14-7