Phenylglyoxal monohydrate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone
161970050 161970250 161971000
ACRO161970050EA 1175 SEK
ACRO161970050 ACRO161970250 ACRO161971000
Phenylglyoxal monohydrate 97%
Phenylglyoxal monohydrate
CAS No.: 1075-06-5
Formel: C₈H₆O₂·H₂O
MW: 152,15 g/mol (134,12 anhy)
Kokpunkt: 142 °C (125 mmHg)
Smältpunkt: 71…76 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149499
CAS nummer: 1075-06-5
Order Now
Specifikation testresultat
Appearance White to light yellow to pink Powder
Infrared spectrum Conforms
Melting point 71 to 76 °C
Titration excess NaOH ≥96.0 %