2,4-Dihydroxypyrimidine-5-carboxylic acid 98%
Leverantör: Thermo Scientific
Synonyms: Isoorotic acid, Uracil-5-carboxylic acid
155200050 155200250
ACRO155200050EA 640 SEK
ACRO155200050 ACRO155200250
2,4-Dihydroxypyrimidine-5-carboxylic acid 98%
2,4-Dihydroxypyrimidine-5-carboxylic acid
Appearance: White to light yellow Crystalline powder
Formel: C₅H₄N₂O₄
MW: 156,10 g/mol
Smältpunkt: 283 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006023
CAS nummer: 23945-44-0
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %