4-Methoxy-3-buten-2-one 90% tech.
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Methoxybut-3-en-2-one
153390100 153390500
ACRO153390100EA 1100 SEK
ACRO153390100 ACRO153390500
4-Methoxy-3-buten-2-one 90% tech.
4-Methoxy-3-buten-2-one
Appearance: Clear yellow Liquid
Formel: C₅H₈O₂
MW: 100,117 g/mol
Kokpunkt: 200 °C (760 mmHg)
Densitet: 0,98 g/cm³
Flampunkt: 63 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008778
CAS nummer: 4652-27-1
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
GC ≥88 %