trans-β-Methylstyrene 97% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: trans-Propenylbenzene, trans-1-Phenyl-1-propene
150150100 150150010 150150500
ACRO150150100EA 2875 SEK
ACRO150150100 ACRO150150010 ACRO150150500
trans-β-Methylstyrene 97% stabilised
trans-beta-Methylstyrene
CAS No.: 873-66-5
Formel: C₉H₁₀
MW: 118,18 g/mol
Kokpunkt: 175 °C (760 mmHg)
Densitet: 0,911 g/cm³
Flampunkt: 58 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009280
CAS nummer: 873-66-5
UN: 3295
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.01 % BHT