3,5-Pyrazoledicarboxylic acid monohydrate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1H-Pyrazole-3,5-dicarboxylic acid hydrate, Pyrazole-3,5-dicarboxylic acid monohydrate, 1H-Pyrazole-3,5-dicarboxylic acid monohydrate
131750050 131750250
ACRO131750050EA 1075 SEK
ACRO131750050 ACRO131750250
3,5-Pyrazoledicarboxylic acid monohydrate 97%
3,5-Pyrazoledicarboxylic acid monohydrate
CAS No.: 3112-31-0
Appearance: White Crystalline powder
Formel: C₅H₄N₂O₄·H₂O
MW: 174,11 g/mol
Smältpunkt: 292…295 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149323
CAS nummer: 303180-11-2
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Water 8.0 to 11.0 %
Titration with NaOH 96.0 to 104.0 %