DL-Homocysteinethiolactone hydrochloride 99%
Leverantör: Thermo Fisher Scientific
Synonyms: DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl
120830250 120831000
ACRO120830250EA 520 SEK
ACRO120830250 ACRO120831000
DL-Homocysteinethiolactone hydrochloride 99%
DL-Homocysteinethiolactone hydrochloride
Appearance: White Crystalline powder
Formel: C₄H₇NOS
MW: 153.62 g/mol
Smältpunkt: 199…203 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012724
CAS nummer: 6038-19-3
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %