Ellagic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,3,7,8-Tetrahydroxychromeno[5,4,3-cde]chromene-5,10-dione, 4,4',5,5',6,6'-Hexahydroxydiphenic acid 2,6,2',6'-dilactone
117740100 117740025 117740500
ACRO117740100EA 2925 SEK
ACRO117740100 ACRO117740025 ACRO117740500
Ellagic acid 97%
Ellagic acid
CAS No.: 476-66-4
Formel: C₁₄H₆O₈
MW: 302,194 g/mol
Smältpunkt: >360 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006914
CAS nummer: 476-66-4
Order Now
Specifikation testresultat
Appearance Beige to green Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
Water ≤0.05 % (Coulometric)
GC ≥95 %
Color scale ≤10 APHA